Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22414F

Sigma-Aldrich

Anti-RSU1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-22414F

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... RSU1(6251)

Immunogen

Immunogen Sequence: GI # 6912638, sequence 1-277
Recombinant ras suppressor protein 1 isoform 1

Application

Anti-RSU1 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Ras suppressor protein 1 is a protein that in humans is encoded by the RSU1 gene and is mapped to human chromosome 10p13. Its expression inhibited the tumorigenesis of a glioblastoma cell line and may inhibit growth regulation in tumors. Rsu-1 may help in inhibiting cell migration by stabilizing the IPP adhesion complex and that Ras activation perturbs this inhibitory function by modulating both Rsu-1 splicing and association of full-length Rsu-1 with IPP. It links the Ras pathway with the IPP complex and the perturbations of cell attachment-dependent signaling that occur in the malignant process.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suryaprabha Chunduru et al.
Journal of neuro-oncology, 60(3), 201-211 (2003-01-04)
Previous studies demonstrated that the Ras suppressor, RSU-1, localizes to human chromosome 10p13, a region frequently deleted in high grade gliomas, and that RSU-1 expression inhibited the tumorigenesis of a glioblastoma cell line. We have now examined RNA from human
Gerard W Dougherty et al.
European journal of cell biology, 87(8-9), 721-734 (2008-04-26)
The link between Ras transformation and enhanced cell migration due to altered integrin signaling is well established in tumorigenesis, however there remain gaps in our understanding of its mechanism. The Ras suppressor, Rsu-1, has recently been linked to the IPP
Human RSU1 is highly homologous to mouse Rsu-1 and localizes to human chromosome 10.
T Tsuda et al.
Genomics, 18(2), 461-462 (1993-11-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service