Immunogen Sequence: GI # 6912602, sequence 100-341
Recombinant ADP-ribosylation factor interacting protein 2 (arfaptin 2)
Application
Anti-ARFIP2 (ab2) antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.
Biochem/physiol Actions
Arfaptin-2 is a protein encoded by the ARFIP2 gene in humans. It binds directly to Rac1 and help in Rac1-mediated signaling pathways. Arfaptin2 contains a Bin/Amphiphysin/Rvs (BAR) domain and directly interacts with proteins of the Arf/Arl family. It is ubiquitously expressed and bound to the GTP and may be involved in Golgi function along with ARF1. Arfaptin-2 is associated in regulating huntingtin protein aggregation, possibly by impairing proteasome function.
Physical form
Solution in phosphate buffered saline containing 0.02% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Arfaptin2 contains a Bin/Amphiphysin/Rvs (BAR) domain and directly interacts with proteins of the Arf/Arl family in their active GTP-bound state. It has been proposed that BAR domains are able to sense membrane curvature and to induce membrane tubulation. We report
The Journal of biological chemistry, 272(9), 5421-5429 (1997-02-28)
ADP-ribosylation factors (ARFs) have been implicated in vesicle transport in the Golgi complex. Employing yeast two-hybrid screening of an HL60 cDNA library using a constitutively active mutant of ARF3 (ARF3.Q71L), as a probe, we have identified a cDNA encoding a
Huntington's disease (HD) is an inherited neurodegenerative disorder. Here we demonstrate that expression of arfaptin 2/POR1 (partner of Rac1) in cultured cells induces the formation of pericentriolar and nuclear aggregates, which morphologically resemble mutant huntingtin aggregates characteristic of HD. Endogenous
The Rac GTP binding proteins are implicated in actin cytoskeleton-membrane interaction in mammalian cells. In fibroblast cells, Rac has been shown to mediate growth factor-induced polymerization of actin to form membrane ruffles and lamellipodia. We report here the isolation of
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.