Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22331B

Sigma-Aldrich

Anti-KAT (ab2) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

manufacturer/tradename

Genway 15-288-22331B

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KAT(10300)

General description

KATNB1 (katanin p80 subunit B 1) forms the regulatory component of katanin protein, which also contains the catalytic p60 subunit. This protein is composed of procon80 domain at its C-terminal, and a WD40 domain at its N-terminal. This gene is highly conserved in humans, spans ~1.6kb, and is composed of four exons. It codes for a protein of 115 amino acids, with a molecular weight of 12.5kDa. It has a wide range of tissue expression such as, thymus, heart, lung, liver, kidney, spleen, colon, skeletal muscle and placenta.

Immunogen

Immunogen Sequence: GI # 5031817, sequence 301-511
Recombinant katanin p80 subunit B 1

Application

Anti-KAT (ab2) antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

The KAT (thiosulfate sulfurtransferase (rhodanese)-like domain containing 1) protein is found around the nuclear membranes and is expressed in several breast, colon and lung carcinoma cell lines, but not in normal breast epithelial cell lines. It is also detected in invasive ductal carcinoma, but not in adjacent tissues and has a role in tumorigenesis.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K P McNally et al.
Journal of cell science, 113 ( Pt 9), 1623-1633 (2000-04-06)
The assembly and function of the mitotic spindle requires the activity of a number of microtubule-binding proteins. Some microtubule-binding proteins bind microtubules in vitro but do not co-localize with microtubules in interphase cells. Instead these proteins associate with specific subregions
Haruka Sudo et al.
Human molecular genetics, 17(16), 2524-2540 (2008-05-21)
Human chromosome region 10q23-24 is one of the most frequently found regions that show loss of heterozygosity in prostate cancers. A candidate tumor suppressor LAPSER1/LZTS2 (LAPSER1) is located in 10q24.3 that has been reported to be deleted as frequently as
Katrin Wenzel et al.
Biological chemistry, 384(5), 763-775 (2003-06-24)
We describe the molecular characterization of a novel human gene on chromosome 1q23.3, termed KAT, which is highly conserved among mammals. The KAT gene spans a genomic region of approximately 1.6 kilobases and consists of 4 exons encoding a 115

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service