Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22317B

Sigma-Aldrich

Anti-SLP-2 (ab2) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$418.00

$418.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$418.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$418.00


Check Cart for Availability

Request a Bulk Order

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

manufacturer/tradename

Genway 15-288-22317B

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLP-2(30968)

General description

The previously assigned protein identifier Q53G29 has been merged into Q9UJZ1. Full details can be found on the UniProt database.

Immunogen

Immunogen Sequence: GI # 7305503, sequence 171-356
Recombinant stomatin (EPB72)-like 2

Application

Anti-SLP-2 (ab2) antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Stomatin-like protein 2 (STOML2) is a member of the stomatin superfamily and is a widely expressed mitochondrial inner membrane protein. It acts as an oncogenic-related protein and is found to be up-regulated in multi-cancers.[1] STOML2 has a role and clinical significance in gastric adenocarcinoma and gallbladder cancer (GBC). Increased expression of STOML2 may lead to unfavorable prognosis for gastric adenocarcinoma patients. STOML2 may play an important role in human GBC tumorigenesis and may serve as a novel prognostic marker in human GBC. It interacts with prohibitin-1 and -2 and binds to the mitochondrial membrane phospholipid cardiolipin. The up-regulation of this gene expression increases cardiolipin content, the formation of metabolically active mitochondrial membranes and also induces mitochondrial biogenesis.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Hong Li et al.
Medical oncology (Northwood, London, England), 31(1), 763-763 (2013-11-22)
Stomatin-like protein 2 (STOML2), a member of the stomatin, has been reported to be upregulated in several human cancers. However, its role and clinical significance in gastric adenocarcinoma remains unclear to date. The purpose of this retrospective study was to
Wei-Xin Wang et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(1), 419-423 (2013-08-07)
Several studies have indicated that overexpression of stomatin-like protein 2 (SLP-2) has been identified in several types of cancer. However, its role and clinical relevance in gallbladder cancer (GBC) is unknown. The purpose of this study was to reveal the
Zhibing Qiu et al.
Journal of cellular biochemistry (2018-10-12)
Previous studies have shown that stomatin-like protein-2 (SLP-2) could regulate mitochondrial biogenesis and function. The study was designed to explore the contribution of SLP-2 to the myocardial ischemia and reperfusion (I/R) injury. Anesthetized rats were treated with SLP-2 and subjected
Darah A Christie et al.
Molecular and cellular biology, 31(18), 3845-3856 (2011-07-13)
Stomatin-like protein 2 (SLP-2) is a widely expressed mitochondrial inner membrane protein of unknown function. Here we show that human SLP-2 interacts with prohibitin-1 and -2 and binds to the mitochondrial membrane phospholipid cardiolipin. Upregulation of SLP-2 expression increases cardiolipin
Wenfeng Cao et al.
PloS one, 8(5), e63890-e63890 (2013-05-15)
Stomatin-like protein 2 (SLP-2), a member of the Stomatin superfamily, has been identified as an oncogenic-related protein and found to be up-regulated in multi-cancers. Nonetheless, the expression pattern and regulation of SLP-2 in human esophageal squamous cell carcinoma (ESCC) remain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service