Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22309F

Sigma-Aldrich

Anti-CDK4 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

manufacturer/tradename

Genway 15-288-22309F

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDK4(1019)

Immunogen

Immunogen Sequence: GI # 4502735, sequence 1-303
Recombinant cyclin-dependent kinase 4

Application

Anti-CDK4 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Cyclin-dependent kinase 4 (also known as cell division protein kinase 4) is an enzyme encoded by the CDK4 gene in humans. It is highly expressed in nasopharyngeal carcinomas (NPC). CDK4 phosphorylates and inactivates retinoblastoma (Rb) protein family members in association with cyclin-D. It mediates progression through the G1- to the S-phase of the cell cycle. Over expression of CDK4 is mostly independent with gene amplification and is a potential prognostic biomarker in NPC and may indicate tumor aggressiveness through cell cycle dysregulation. It is the main molecular feature of well-differentiated liposarcomas (WDLS) and dedifferentiated liposarcomas (DDLS). Inhibition of CDK4 partly restores G(1)-S arrest and sensitizes cells to doxorubicin-mediated cell death in MYCN-amplified cells with an intact p53 pathway. CDK4 up-regulation takes place in oral cancer progression.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sina Gogolin et al.
Cell cycle (Georgetown, Tex.), 12(7), 1091-1104 (2013-03-07)
Relapse with drug-resistant disease is the main cause of death in MYCN-amplified neuroblastoma patients. MYCN-amplified neuroblastoma cells in vitro are characterized by a failure to arrest at the G(1)-S checkpoint after irradiation- or drug-induced DNA damage. We show that several
Seung Eun Lee et al.
Histology and histopathology, 29(1), 127-138 (2013-07-16)
The amplification of MDM2 and CDK4 is the main molecular feature of well-differentiated liposarcomas (WDLS) and dedifferentiated liposarcomas (DDLS). Although the diagnostic usefulness of this molecular characteristic in liposarcomas has been investigated, its prognostic utility of quantitative gene level has
Ming J Poi et al.
Anticancer research, 34(7), 3285-3292 (2014-07-02)
While aberrant expression of cyclin-dependent kinase-4 (CDK4) has been found in squamous cell carcinoma of the head and neck (SCCHN), the associations between CDK4 and its regulators, namely, cyclin D1, cyclin E, gankyrin, SEI1, and BMI1 in gene expression remain
Tzu-Ju Chen et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(7), 7209-7216 (2014-04-29)
Data mining in the public domain demonstrates that cyclin-dependent kinase 4 (CDK4) is highly expressed in nasopharyngeal carcinomas (NPC). Associated with cyclin-D, CDK4 phosphorylates and inactivates retinoblastoma (Rb) protein family members and mediates progression through the G1- to the S-phase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service