Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22293

Sigma-Aldrich

Anti-SNAPAP antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

manufacturer/tradename

Genway 15-288-22293

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNAPAP(23557)

Immunogen

Immunogen Sequence: GI # 6912674, sequence 31-136
Recombinant SNAP-associated protein

Application

Anti-SNAPAP antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

SNARE-associated protein Snapin is a protein encoded by the SNAPIN gene in humans. It is a 15kDa protein present in neuronal and non-neuronal cells and is implicated in the regulation of exocytosis and endocytosis. Synaptic vesicle docking and fusion are mediated by the assembly of a stable SNARE core complex of proteins. It includes the synaptic vesicle membrane protein VAMP/synaptobrevin and the plasmalemmal proteins syntaxin and SNAP-25. Snapin is an important component of the neurotransmitter release process through its modulation of the sequential interactions between the SNAREs and synaptotagmin.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Ilardi et al.
Nature neuroscience, 2(2), 119-124 (1999-04-09)
Synaptic vesicle docking and fusion are mediated by the assembly of a stable SNARE core complex of proteins, which include the synaptic vesicle membrane protein VAMP/synaptobrevin and the plasmalemmal proteins syntaxin and SNAP-25. We have now identified another SNAP-25-binding protein
Aaron Navarro et al.
Biochemistry, 51(16), 3470-3484 (2012-04-05)
Snapin is a 15 kDa protein present in neuronal and non-neuronal cells that has been implicated in the regulation of exocytosis and endocytosis. Protein kinase A (PKA) phosphorylates Snapin at Ser-50, modulating its function. Likewise, mutation of Cys-66, which mediates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service