Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22289F

Sigma-Aldrich

Anti-WDR4 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-22289F

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WDR4(10785)

General description

WDR4 (WD repeat domain 4) is a member of the WD repeat protein family, and the gene is localized to human chromosome 21q22.3. This gene is composed of 11 exons, and spans 37kb. Alternative splicing results in two RNA variants of sizes 1.5kb and 2.1kb, where the smaller one is predominantly expressed in fetal tissues. This protein contains four guanine nucleotide-binding WD repeats, and is composed of 412 amino acids. It is the human homologue of TRM82 (tRNA methylase), a protein found in Saccharomyces cerevisiae.

Immunogen

Immunogen Sequence: GI # 16445432, sequence 1-266
Recombinant WD repeat domain 4 protein isoform 2

Application

Anti-WDR4 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

WDR4 (WD repeat domain 4) is mapped between PDE9A and NDUFV3, a region where several genetic disorders are mapped. It forms a protein complex along with Trm8. WDR4 is required for m7G-methyltransferase activity.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Michaud et al.
Genomics, 68(1), 71-79 (2000-08-22)
To identify candidate genes for Down syndrome phenotypes or disorders that map to human chromosome 21q22.3, trapped exons are being used to isolate full-length transcripts. We isolated a full-length cDNA (WDR4) encoding a novel WD-repeat protein and its mouse homologue.
Robert A Cartlidge et al.
The EMBO journal, 24(9), 1696-1705 (2005-04-30)
A substrate for protein kinase B (PKB)alpha in HeLa cell extracts was identified as methyltransferase-like protein-1 (METTL1), the orthologue of trm8, which catalyses the 7-methylguanosine modification of tRNA in Saccharomyces cerevisiae. PKB and ribosomal S6 kinase (RSK) both phosphorylated METTL1
Andrei Alexandrov et al.
RNA (New York, N.Y.), 8(10), 1253-1266 (2002-10-31)
7-methylguanosine (m7G) modification of tRNA occurs widely in eukaryotes and bacteria, is nearly always found at position 46, and is one of the few modifications that confers a positive charge to the base. Screening of a Saccharomyces cerevisiae genomic library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service