Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22011

Sigma-Aldrich

Anti-GADD45G antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Growth arrest and DNA-damage-inducible protein GADD45 γ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

manufacturer/tradename

Genway 15-288-22011

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GADD45G(10912)

General description

The gene is mapped to human chromosome 9q22.1-q22.2 and contains a 477-bp open reading frame that encodes a protein of 159 amino acid residues. It is found to be expressed in heart, placenta, liver, skeletal muscle, prostate, testis, and ovary.[1] The active form of GADD45G (growth arrest and DNA-damage-inducible, γ) is a homodimer and its dimerization is essential for apoptosis and growth inhibition.[2]

Immunogen

Immunogen Sequence: GI # 5729836, sequence 1-159
Recombinant growth arrest and DNA-damage-inducible, gamma

Biochem/physiol Actions

GADD45G (growth arrest and DNA-damage-inducible, γ) gene encodes a protein that is expressed in increased levels under stress, growth arrest conditions and treatment with DNA-damaging agents such as UV (ultraviolet) and γ-irradiation. It mediates activation of MTK1/MEKK4 kinase (mitogen-activated protein kinase kinase kinase 4) upon environmental stress, which in turn activates p38 and JNK MAPK (c-Jun N-terminal kinase/ mitogen-activated protein kinase) pathways that regulate cell cycle and apoptosis.[3] The encoded protein functions in DNA repair, negative growth control, genomic stability, cell cycle checkpoints and apoptosis.[2]

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular cloning, expression, and mapping of a novel human cDNA, GRP17, highly homologous to human gadd45 and murine MyD118.
Suzuki M
Journal of Human Genetics, 44, 300-303 (1999)
Wenzheng Zhang et al.
Protein & cell, 2(10), 814-826 (2011-11-08)
The human Gadd45 protein family plays critical roles in DNA repair, negative growth control, genomic stability, cell cycle checkpoints and apoptosis. Here we report the crystal structure of human Gadd45γ [corrected], revealing a unique dimer formed via a bundle of
M Takekawa et al.
Cell, 95(4), 521-530 (1998-11-25)
The stress-responsive p38 and JNK MAPK pathways regulate cell cycle and apoptosis. A human MAPKKK, MTK1 (= MEKK4), mediates activation of both p38 and JNK in response to environmental stresses. Using a yeast two-hybrid method, three related proteins, GADD45alpha (=

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service