Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW21974B

Sigma-Aldrich

Anti-RTSβ (ab2) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-RTS 946;

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21974B

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... rTS(55556)

Immunogen

Immunogen Sequence: GI # 8923791, sequence 345-416
Recombinant RTS β protein

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service