Immunogen Sequence: GI # 18641347, sequence 482-552
Recombinant protein tyrosine phosphatase, receptor type, C isoform 1
Application
Anti-PTPRC antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.
Biochem/physiol Actions
Protein tyrosine phosphatase, receptor type, C is an enzyme encoded by the PTPRC gene in humans. It is also known as CD45 antigen (CD stands for cluster of differentiation) and is originally called leukocyte common antigen (LCA). PTPRC acts as a binding receptor for galectin-3 and helps in inducing apoptosis of Jurkat cells. It plays a novel role on innate immune cells in promoting lymphopenia-induced T cell proliferation. The leukocyte-specific tyrosine phosphatase impacts T cell development and activation by modulating TCR signaling.
Physical form
Solution in phosphate buffered saline containing 0.02% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Galectin-3 has been reported to induce apoptosis of Jurkat cells through binding receptors such as CD45. CD45RABC is heavily O-glycosylated and N-glycosylated, while CD45RO is only N-glycosylated. In this study, no apoptosis induced by galectin-3 was detected in CD45RO-transfected cells
Journal of immunology (Baltimore, Md. : 1950), 193(6), 2831-2842 (2014-08-13)
The leukocyte-specific tyrosine phosphatase, CD45, severely impacts T cell development and activation by modulating TCR signaling. CD45-deficient (CD45KO) mice have reduced peripheral T cell numbers where CD8 T cells are underrepresented. In this article, we show that CD45KO mice are
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.