Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW21429

Sigma-Aldrich

Anti-ADAM23 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-A disintegrin and metalloproteinase domain 23

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$418.00

$418.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
50 μG
$418.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$418.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21429

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADAM23(8745)

Immunogen

Immunogen Sequence: GI # 4501913, sequence 72-144
Recombinant ADAM metallopeptidase domain 23 preproprotein

Application

Anti-ADAM23 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Disintegrin and metalloproteinase domain-containing protein 23 is an enzyme encoded by the ADAM23 gene in humans and is a member of a disintegrin and metalloprotease (ADAM) family. It is higly expressed in the brain and may function as integrin ligands in the brain along with other proteins. ADAM23 is associated with the progression of non-small-cell lung carcinoma (NSCLC) and its downregulation may be correlated with promoter methylation. It may be down-regulated by aberrant promoter hypermethylation during the progression of colorectal cancer. ADAM23 is down-regulated by methylation in brain tumors, which is a rare event.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Sagane et al.
The Biochemical journal, 334 ( Pt 1), 93-98 (1998-08-07)
Cellular disintegrins are a family of membrane-anchored proteins structurally related to snake venom disintegrins, and are potential regulators of cell-cell and cell-matrix interactions. The members of this protein family are also called ADAMs (a disintegrin and metalloproteinase) or MDC proteins
Jin-Sung Choi et al.
International journal of cancer, 124(6), 1258-1262 (2008-12-18)
Promoter hypermethylation of the ADAM23 gene, which is normally involved in cell-to-cell and cell-to matrix adhesion, has been reported in pancreatic, breast and brain cancer, and recently the role of this gene was examined in gastric cancer. In this study
Chunyan Hu et al.
International journal of experimental pathology, 92(5), 333-339 (2011-03-25)
ADAM23, a member of a disintegrin and metalloprotease (ADAM) family, has been reported to be expressed in several types of tumours. The exact role of ADAM23 and the possible mechanisms in which it is involved in non-small-cell lung carcinoma (NSCLC)
Fabrício F Costa et al.
Neuroscience letters, 380(3), 260-264 (2005-05-03)
The ADAMs comprises a family of cell surface proteins with putative roles in cell-cell and/or cell-matrix interactions and in protease activities. In this work, we have examined the expression level and the methylation status of the 5' upstream region of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service