Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW21421

Sigma-Aldrich

Anti-MITF antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Microphthalmia-associated transcription factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MITF(4286)

General description

MITF (microphthalmia-associated transcription factor) is a b-HLH-Zip (basic helix-loop-helix leucine zipper) transcription factor, which is specific to melanocytes. It is a member of the Myc superfamily, and the gene is localized to human chromosome 3. It is 229kb in size. It belongs to the MiT (microphthalmia) family of transcription factors, which also includes TFEB (transcription factor EB), TFEC and TFE3. Due to alternative splicing, this protein is produced as multiple different isoforms such as TF-A, MITF-B, MITF-C, MITF-D, MITF-E, MITF-H, MITF-J, MITF-M and MITF-Mc. The N-termini of these isoforms vary, and they have different tissue expression patterns. The shortest isoform is MITF-M, which is only expressed in melanoma cells and melanocytes. Its transcriptional activity is modulated by its two TADs (transactivation domains), present at N- and C-termini.

Immunogen

Immunogen Sequence: GI # 4557755, sequence 287-417
Recombinant microphthalmia-associated transcription factor isoform 4

Application

Anti-MITF antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Microphthalmia-associated transcription factor (MITF) is involved in lineage-specific pathway regulation of many types of cells. Mitf is required for the proper development of osteoclasts, melanocytes, retinal pigment epithelial cells, mast cells and natural killer cells. It can be a valuable adjunct in the diagnosis of metastatic tumors that are suspicious for melanoma but negative for other melanoma markers.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruifeng Guo et al.
International journal of clinical and experimental pathology, 6(8), 1658-1664 (2013-08-08)
Metastatic malignant melanoma has a wide spectrum of histopathologic patterns and often lacks melanin pigment. Without a known primary tumor, the diagnosis of metastatic malignant melanoma relies on a combination of morphology and immunohistochemical profile. Infrequently, commonly used markers for
Mariusz L Hartman et al.
Cellular and molecular life sciences : CMLS, 72(7), 1249-1260 (2014-12-01)
MITF (microphthalmia-associated transcription factor) represents a melanocytic lineage-specific transcription factor whose role is profoundly extended in malignant melanoma. Over the last few years, the function of MITF has been tightly connected to plasticity of melanoma cells. MITF participates in executing
Christine L Hershey et al.
Bone, 34(4), 689-696 (2004-03-31)
The Microphthalmia-associated transcription factor (Mitf) is required for the proper development of several cell lineages including osteoclasts, melanocytes, retinal pigment epithelial cells, mast cells and natural killer cells. Mutations in Mitf in multiple organisms result in osteopetrosis due to defective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service