Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW21263

Sigma-Aldrich

Anti-TUBB2A antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Tubulin β-2 chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

manufacturer/tradename

Genway 15-288-21263

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUBB(203068)

Immunogen

Immunogen Sequence: GI # 29788785, sequence 356-427
Recombinant tubulin, β polypeptide

Application

Anti-TUBB2A antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Tubulin β chain is a protein encoded by the TUBB gene in humans and is mapped to the HLA class I region at 6p21.3. Mutation in TUBB gene may cause certain forms of HLA-linked microtubule dysfunction, including immotile cilia syndrome.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Volz et al.
Human genetics, 93(1), 42-46 (1994-01-01)
An expressed beta-tubulin gene (TUBB) has previously been localized to chromosome region 6pter-p21 in man. By using a panel of deletion mutant cell lines and radiation-reduced hybrids containing fragments of chromosome 6, the TUBB locus could be mapped to the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service