Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW21249

Sigma-Aldrich

Anti-C13orf12 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Chromosome 13 open reading frame 12, Anti-HSPC036 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

manufacturer/tradename

Genway 15-288-21249

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

C13orf12, also called POMP (proteasome maturation protein), is a chaperone protein for proteasome, and has a molecular weight of 16kDa. It is thought to exist as a tetramer with the molecular weight of 64kDa. It is both a cytosolic and a nuclear protein. It has a ubiquitous expression pattern, and the corresponding gene is localized to human chromosome 13q.

Immunogen

Immunogen Sequence: GI # 7705429, sequence 1-141
Recombinant proteasome maturation protein

Application

Anti-C13orf12 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

POMP (proteasome maturation protein), or C13orf12, acts as a chaperone which is essential for the maturation of the 20S proteasome proteolytic core complex. It has an important role in the terminal differentiation of human epidermis. Deletion in this gene is linked with KLICK syndrome, which is a rare autosomal-recessive skin disorder. The deletion results in modified distribution of proteasome in the epidermis, causing altered formation of outer layers of skin.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johanna Dahlqvist et al.
American journal of human genetics, 86(4), 596-603 (2010-03-17)
KLICK syndrome is a rare autosomal-recessive skin disorder characterized by palmoplantar keratoderma, linear hyperkeratotic papules, and ichthyosiform scaling. In order to establish the genetic cause of this disorder, we collected DNA samples from eight European probands. Using high-density genome-wide SNP
Melanie M Hoefer et al.
International journal of biological macromolecules, 38(3-5), 259-267 (2006-04-21)
The proteasome is a multisubunit complex with a central role in non-lysosomal proteolysis and the processing of proteins for presentation by the MHC class I pathway. The 16kDa proteasome maturation protein POMP (also named proteassemblin or hUmp1) acts as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service