Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW21183

Sigma-Aldrich

Anti-BRCA1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Breast and ovarian cancer susceptibility protein splice variant

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21183

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BRCA1(672)

Immunogen

Immunogen Sequence: GI # 6552299, sequence 1395-1454
Recombinant breast cancer 1, early onset isoform 1

application

Anti-BRCA1 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

BRCA1 is a human tumor suppressor gene. Its main function is to maintain genomic stability through the assembly of multiple protein complexes involved in DNA repair, cell-cycle arrest and transcriptional regulation. BRCA1 is associated with breast and ovarian tumor suppression. Mutations in BRCA1 is involved in the pathogenesis of breast and ovarian cancer. BRCA1 promoter methylation is associated with better response to platinum-taxane-based therapy in epithelial ovarian cancer (EOC). BRCA1 mutation may drive the formation of tumors in estrogen-regulated tissues.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Ruscito et al.
European journal of cancer (Oxford, England : 1990), 50(12), 2090-2098 (2014-06-04)
Mutations in BRCA1/2 genes are involved in the pathogenesis of breast and ovarian cancer. Inactivation of these genes can also be mediated by hypermethylation of CpGs in the promoter regions. Aim of this study was to analyse the clinical impact
Kienan I Savage et al.
Cancer research, 74(10), 2773-2784 (2014-03-19)
Germline mutations in BRCA1 predispose carriers to a high incidence of breast and ovarian cancers. BRCA1 functions to maintain genomic stability through critical roles in DNA repair, cell-cycle arrest, and transcriptional control. A major question has been why BRCA1 loss
Sarah J Hill et al.
Genes & development, 28(17), 1957-1975 (2014-09-04)
BRCA1 is a breast and ovarian tumor suppressor. Given its numerous incompletely understood functions and the possibility that more exist, we performed complementary systematic screens in search of new BRCA1 protein-interacting partners. New BRCA1 functions and/or a better understanding of
T Ignatov et al.
Journal of cancer research and clinical oncology, 140(9), 1457-1463 (2014-05-16)
The aim of the current study was to investigate the role of BRCA1 promoter methylation as predictive factor of response to platinum-taxane-based therapy in sporadic ovarian cancer. BRCA1 promoter methylation was analyzed in 42 sporadic epithelial ovarian cancers. The results
Kienan I Savage et al.
Molecular cell, 54(3), 445-459 (2014-04-22)
Mutations within BRCA1 predispose carriers to a high risk of breast and ovarian cancers. BRCA1 functions to maintain genomic stability through the assembly of multiple protein complexes involved in DNA repair, cell-cycle arrest, and transcriptional regulation. Here, we report the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service