Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW21182

Sigma-Aldrich

Anti-ESR1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ER, Anti-Estrogen receptor 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

manufacturer/tradename

Genway 15-288-21182

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ESR1(2099)

Immunogen

Immunogen Sequence: GI # 4503603, sequence 255-367
Recombinant estrogen receptor 1

Application

Anti-ESR1 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

ERα is encoded by the ESR1 (estrogen receptor 1) gene in humans and is mapped to chmosome 6q25.1. It may play an important role in the development and progression of cerebral infarction. ESR1 genetic polymorphisms may contribute to the development of cerebral infarction. Methylation of ESR1 influences ovarian cancer development and may play a role regarding prognosis of ovarian carcinoma. It may be associated with tumorigenesis of breast cancer.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Verena Kirn et al.
Journal of cancer research and clinical oncology, 140(10), 1681-1687 (2014-06-09)
Ovarian carcinoma is the third most common gynecological cancer and only short recurrence-free survival and overall survival times are achieved. The role of the estrogen receptor expression is well studied in breast cancer and breast cancer cell lines. Patients with
Hong-Hua Gao et al.
DNA and cell biology, 33(9), 605-615 (2014-04-30)
A number of studies have documented that estrogen receptor α (ESR1) may play an important role in the development and progression of cerebral infarction, but many existing studies have yielded inconclusive results. This meta-analysis was performed to evaluate the relationships
Jim-Ray Chen et al.
Virchows Archiv : an international journal of pathology, 464(6), 689-699 (2014-04-24)
Immunohistochemical expression of ERα, encoded by the ESR1 (estrogen receptor 1) gene located at 6q25.1, is the most important determinant of responsiveness to endocrine therapy in breast cancer. The prevalence and significance of ESR1 amplification in breast cancer remain controversial.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service