Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW21167

Sigma-Aldrich

Anti-NFKB1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-NFKB-p105, Anti-Nuclear factor NF-kappa-B p105 subunit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

manufacturer/tradename

Genway 15-288-21167

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFKB1(4790)

Immunogen

Immunogen Sequence: GI # 10835177, sequence 432-499
Recombinant nuclear factor of kappa light polypeptide gene enhancer in B-cells 1 (p105)

Application

Anti-NFKB1 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Nuclear factor NF-κ-B p105 subunit is a protein encoded by the NFKB1 gene in humans. It is a DNA binding subunit of B-cell protein that interacts with the immunoglobulin κ light-chain gene enhancer. Tumor necrosis factor α (TNF α) helps in activation and induces expression of NFKB1 gene. Oxaliplatin reactivates HIV-1 expression by inducing nuclear factor κ B (NF-κB) nuclear translocation and may be a potential drug candidate for anti-latency therapies.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Meyer et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(3), 966-970 (1991-02-01)
The DNA binding subunit of nuclear factor kappa B (NF-kappa B), a B-cell protein that interacts with the immunoglobulin kappa light-chain gene enhancer, has been purified from nuclei of human HL-60 cells stimulated with tumor necrosis factor alpha (TNF alpha)
Xiaoli Zhu et al.
Biochemical and biophysical research communications, 450(1), 202-207 (2014-06-01)
Reactivation of latent HIV-1 is a promising strategy for the clearance of the viral reservoirs. Because of the limitations of current agents, identification of new latency activators is urgently required. Using an established model of HIV-1 latency, we examined the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service