Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW21160

Sigma-Aldrich

Anti-MDM2 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Mouse double minute 2 homolog isoform MDM2, Anti-Ubiquitin-protein ligase E3 Mdm2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

manufacturer/tradename

Genway 15-288-21160

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MDM2(4193)

Immunogen

Immunogen Sequence: GI # 4505137, sequence 389-491
Recombinant mouse double minute 2 homolog isoform MDM2

Application

Anti-MDM2 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Mouse double minute 2 homolog (MDM2) is a protein encoded by the MDM2 gene in humans. It is a p53 target gene. It is an E3 ubiquitin ligase. MDM2 is a key negative regulator of p53 protein and it forms an auto-regulatory feedback loop with p53. It is an oncogene with both p53-dependent and p53-independent oncogenic activities. MDM2 is highly regulated at the transcriptional, translational and post-translational levels. MDM2 dysregulation impacts significantly upon the p53 functions and in turn, tumorigenesis. It is one of the strong candidates for cellular proteins. MDM2 is associated with increased risk and aggressiveness of neuroblastoma and other tumor entities.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuhan Zhao et al.
Acta biochimica et biophysica Sinica, 46(3), 180-189 (2014-01-07)
Tumor suppressor p53 plays a central role in preventing tumor formation. The levels and activity of p53 is under tight regulation to ensure its proper function. Murine double minute 2 (MDM2), a p53 target gene, is an E3 ubiquitin ligase.
Ali Rihani et al.
Pediatric blood & cancer, 61(10), 1867-1870 (2014-01-07)
While a polymorphism located within the promoter region of the MDM2 proto-oncogene, SNP309 (T > G), has previously been associated with increased risk and aggressiveness of neuroblastoma and other tumor entities, a protective effect has also been reported in certain other cancers.
J D Oliner et al.
Nature, 358(6381), 80-83 (1992-07-02)
Despite extensive data linking mutations in the p53 gene to human tumorigenesis, little is known about the cellular regulators and mediators of p53 function. MDM2 is a strong candidate for one such cellular protein; the MDM2 gene was originally identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service