Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW21125

Sigma-Aldrich

Anti-MMP14 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-MT1-MMP, Anti-Matrix metalloproteinase 14

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human

manufacturer/tradename

Genway 15-288-21125

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP14(4323)

Immunogen

Immunogen Sequence: GI # 4826834, sequence 126-230
Recombinant matrix metalloproteinase 14 preproprotein

Application

Anti-MMP14 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Matrix metalloproteinase-14 is an enzyme encoded by the MMP14 gene in humans. It is an important member of the matrix metalloproteinase (MMP) family. It is a degrading enzyme having complex and specific regulation networks. MMP-14 facilitates local extracellular matrix (ECM) degradation and acts as a major mediator of cell migration, angiogenesis and invasion. It is involved in the breakdown of extracellular matrix in normal physiological processes, such as mediating cell invasion through the extracellular matrix (ECM).[1] Knockdown of this gene results in increase of N-cadherin adhesions, which are prerequisite for fibronectin matrix formation. MMP-14 signals autophagy, a process which may contribute to the chemoresistance phenotype in brain cancer cells. Mutation in MMP14 causes an autosomal-recessive, multicentric disorder, winchester syndrome, with multicentric osteolysis and arthritis.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jonathan Pratt et al.
Cellular signalling, 26(5), 917-924 (2014-01-28)
Plant lectins have been considered as possible anti-tumor drugs because of their property to induce autophagic cell death. Given that expression of membrane type-1 matrix metalloproteinase (MT1-MMP) has been found to regulate expression of the autophagy biomarker Bcl-2/adenovirus E1B 19kDa
Brad R Evans et al.
American journal of human genetics, 91(3), 572-576 (2012-08-28)
The "vanishing bone" syndromes represent a group of rare skeletal disorders characterized by osteolysis and joint destruction, which can mimic severe rheumatoid arthritis. Winchester syndrome was one of the first recognized autosomal-recessive, multicentric forms of the disorder. It was originally
Takahisa Takino et al.
Biochemical and biophysical research communications, 450(2), 1016-1020 (2014-07-01)
Fibronectin matrix formation requires the increased cytoskeletal tension generated by cadherin adhesions, and is suppressed by membrane-type 1 matrix metalloproteinase (MT1-MMP). In a co-culture of Rat1 fibroblasts and MT1-MMP-silenced HT1080 cells, fibronectin fibrils extended from Rat1 to cell-matrix adhesions in
Amanda Haage et al.
Biochemical and biophysical research communications, 450(1), 213-218 (2014-06-01)
Matrix metalloproteinases (MMPs) are extracellular matrix (ECM) degrading enzymes and have complex and specific regulation networks. This includes activation interactions, where one MMP family member activates another. ECM degradation and MMP activation can be initiated by several different stimuli including
Dane K Lund et al.
American journal of physiology. Cell physiology, 307(2), C140-C149 (2014-06-06)
The twenty-five known matrix metalloproteases (MMPs) and their endogenous inhibitors, tissue inhibitors of metalloproteases (TIMPs), mediate cell invasion through the extracellular matrix (ECM). In a comparative three-dimensional assay, we analyzed human and mouse satellite cells' competence to invade an artificial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service