Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW21102

Sigma-Aldrich

Anti-CXCL13 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-SCYB13, Anti-Small inducible cytokine B13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21102

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CXCL13(10563)

Immunogen

Immunogen Sequence: GI # 5453577, sequence 65-94
Recombinant chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant)

Application

Anti-CXCL13 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

C-X-C motif chemokine 13 (CXCL13) is a protein (B-cell chemokine) encoded by the CXCL13 gene in humans. It functions as a ligand and may guide B lymphocytes to follicles in secondary lymphoid organs. CXCL13 mediates B-cell trafficking and get increased proportionately to disease activity, in many antibody-mediated syndromes. Its level is increased and is a prognostic biomarker in patients with idiopathic pulmonary fibrosis (IPF) and COPD. CXCL13 concentration is found to be significantly elevated in the cerebrospinal fluid (CSF) of the group with aseptic meningoencephalitis compared with the group with aseptic meningitis. It may act as a useful marker for predicting the prognosis of aseptic meningitis. CXCL13 may also act as an a biomarker in multiple sclerosis.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enrique Alvarez et al.
Multiple sclerosis (Houndmills, Basingstoke, England), 19(9), 1204-1208 (2013-01-17)
CXCL13, a B-cell chemokine, has been proposed as a biomarker in a variety of conditions, some of which can mimic multiple sclerosis and can have very high levels. In this case-control study, cerebrospinal fluid (CSF) CXCL13 was elevated in multiple
Juichi Fujimori et al.
Journal of neuroimmunology, 273(1-2), 77-84 (2014-06-09)
In exceptional cases, patients with aseptic meningitis eventually develop aseptic meningoencephalitis. To find a candidate marker for the development of aseptic meningoencephalitis in adult patients diagnosed with aseptic meningitis, we compared 12 different cytokines/chemokines in cerebrospinal fluid (CSF) from 5
Louis J Vuga et al.
American journal of respiratory and critical care medicine, 189(8), 966-974 (2014-03-19)
C-X-C motif chemokine 13 (CXCL13) mediates B-cell trafficking and is increased, proportionately to disease activity, in many antibody-mediated syndromes. Dysregulated B cells have recently been implicated in idiopathic pulmonary fibrosis (IPF) pathogenesis. To determine if CXCL13 is associated with IPF
M D Gunn et al.
Nature, 391(6669), 799-803 (1998-03-05)
Secondary lymphoid organs (spleen, lymph nodes and Peyer's patches) are divided into compartments, such as B-cell zones (follicles) and T-cell zones, which provide specialized environments for specific steps of the immune response. Migration of lymphocyte subsets into these compartments is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service