LTBR (lymphotoxin β receptor) is a member of the tumor necrosis factor (TNF) superfamily, which also includes LTα and LTβ. It is expressed on stromal, parenchymal and myeloid cells. It is a part of the noncanonical/canonical nuclear factor-κB (NF-κB) signaling pathway.
Immunogen
Immunogen Sequence: GI # 4505039, sequence 106-159
Recombinant lymphotoxin β receptor
Application
Anti-LTBR antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.
Biochem/physiol Actions
Lymphotoxin β receptor signaling serves pleiotropic functions including the control of lymphoid organ development and support of efficient immune responses against pathogens. LTBR signaling induces chemokine, cytokine or adhesion molecule expression, cell proliferation and cell survival. LTBR is important for development and organization of the secondary lymphoid tissues and its polymorphisms may be associated with risk of IgA nephropathy (IgAN).
Physical form
Solution in phosphate buffered saline containing 0.02% sodium azide.
Lymphotoxin beta receptor (LTBR) is essential for development and organization of the secondary lymphoid tissues. To investigate whether LTBR polymorphisms are associated with IgA nephropathy (IgAN) in Korean children, One hundred ninety nine patients with IgAN and 289 controls were
The cytokines lymphotoxin (LT) alpha, beta and their receptor (LTbetaR) belong to the tumor necrosis factor (TNF) superfamily, whose founder-TNFalpha-was initially discovered due to its tumor necrotizing activity. LTbetaR signaling serves pleiotropic functions including the control of lymphoid organ development
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.