Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW21051

Sigma-Aldrich

Anti-CX3CR1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CX3C chemokine receptor 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21051

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CX3CR1(1524)

General description

CX3CR1 (chemokine (C-X3-C motif) receptor 1) is a surface receptor for chemoattractant cytokines. It is expressed by circulating monocytes, tissue macrophages and dendritic cells, T cells and natural killer (NK) cells. It acts as a receptor for CX3CL1 chemokine.[1] In brain, it is expressed only in microglial cells.

Immunogen

Immunogen Sequence: GI # 4503171, sequence 1-21
Recombinant chemokine (C-X3-C motif) receptor 1

Application

Anti-CX3CR1 antibody produced in chicken is suitable for indirect ELISA and western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

CX3C chemokine receptor 1 (CX3CR1) plays an important role in modulating inflammatory responses that includes monocyte homeostasis and macrophage phenotype and function. CX3CR1 is associated with angiogenic macrophage survival in the tumor microenvironment contributing to tumor metastasis.[1] It is implicated as a candidate gene for age-related macular degeneration (AMD). CX3CR1 polymorphisms may influence the incidence and the outcome of acute myocardial infarction.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Pucci et al.
BioMed research international, 2013, 451349-451349 (2013-12-07)
Fractalkine is a proinflammatory chemokine that participates in atherosclerotic process mediating the interactions of vascular cells and leukocytes and selective recruitment of Th1 lymphocytes, through interaction with CX3CR1 receptor. The polymorphism of the fractalkine receptor 280M-containing haplotype, which codifies for
Jiao Zheng et al.
Molecular cancer, 12(1), 141-141 (2013-11-20)
Macrophages, the key component of the tumor microenvironment, are differentiated mononuclear phagocyte lineage cells that are characterized by specific phenotypic characteristics that have been implicated in tumor growth, angiogenesis, and invasion. CX3CR1, the chemoattractant cytokine CX3CL1 receptor, plays an important
Alan Lopez-Lopez et al.
PloS one, 9(5), e96528-e96528 (2014-05-09)
The objective of this study was to investigate the association of functional variants of the human CX3CR1 gene (Fractalkine receptor) with the risk of Amyotrophic Lateral Sclerosis (ALS), the survival and the progression rate of the disease symptoms in a
Debra A Schaumberg et al.
JAMA ophthalmology, 132(1), 84-95 (2013-11-30)
The CX3CR1 gene is implicated as a candidate gene for age-related macular degeneration (AMD) through several lines of evidence. There is uncertainty, however, as to whether common genetic variants in CX3CR1 alter risk of AMD, since prior studies have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service