Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW21034

Sigma-Aldrich

Anti-Interleukin-21 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-IL-21

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21034

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL21(59067)

Immunogen

Immunogen Sequence: GI # 46575632, sequence 33-111
Recombinant interleukin 21

Application

Anti-Interleukin-21 antibody produced in chicken is suitable for indirect ELISA and western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Interleukin-21 (IL-21) is a protein encoded by the IL21 gene in humans. It is a cytokine that has potent regulatory effects on cells of the immune system, including natural killer (NK) cells and cytotoxic T cells. Cytokines are crutial for the regulation of haematopoiesis and immune responses. It can influence lymphocyte development. IL21 has an important role in the proliferation and maturation of natural killer (NK) cell populations from bone marrow. The gene may play a role in the development of Systemic lupus erythematosus (SLE). IL-21 is essential for B cell activation, expansion and PC differentiation induced by direct interaction of B cells with activated T cells. Polymorphisms of this gene is associated with several cancers.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Lan et al.
Gene, 538(1), 94-98 (2014-01-18)
Systemic lupus erythematosus (SLE) is one of the common autoimmune diseases, with complex genetic components. Interleukin-21 (IL-21) is the most recently discovered member of the type-I cytokine family, which has a variety of effects on the immune system, including B
Mang Xiao et al.
Gene, 537(1), 15-19 (2014-01-07)
Polymorphisms in Interleukin (IL)-21 have been researched in several cancers, but the association between IL-21 polymorphisms and thyroid cancer remains unclarified. This case-control study explored the role of five tagSNPs (rs12508721C>T, rs907715G>A, rs13143866G>A, rs2221903A>G and rs4833837A>G) in IL-21 gene in
Stefan Kuchen et al.
Journal of immunology (Baltimore, Md. : 1950), 179(9), 5886-5896 (2007-10-20)
During T cell-B cell collaboration, plasma cell (PC) differentiation and Ig production are known to require T cell-derived soluble factors. However, the exact nature of the cytokines produced by activated T cells that costimulate PC differentiation is not clear. Previously
J Parrish-Novak et al.
Nature, 408(6808), 57-63 (2000-11-18)
Cytokines are important in the regulation of haematopoiesis and immune responses, and can influence lymphocyte development. Here we have identified a class I cytokine receptor that is selectively expressed in lymphoid tissues and is capable of signal transduction. The full-length

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service