Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW21016

Sigma-Aldrich

Anti-PCOLN3 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Charged multivesicular body protein 1a, Anti-KIAA0047, Anti-Vacuolar protein sorting 46-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21016

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHMP1A(5119)

Immunogen

Immunogen Sequence: GI # 9885433, sequence 80-196
Recombinant charged multivesicular body protein 1/chromatin modifying protein 1

Application

Anti-PCOLN3 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Charged multivesicular body protein 1a is a protein encoded by the CHMP1A gene in humans and is a member of the ′Endosomal Sorting Complex Required for Transport′ (ESCRT-III) family. It plays a role in the cytoplasm in sorting proteins to the multivesicular body (MVB) and functions as a tumor suppressor gene in renal cells. It may be a useful target for treatment of renal cell carcinoma (RCC). Overexpression of this gene induces growth inhibition, chromatin condensation and p53 phosphorylation. The gene regulates Ataxia telangiectasia mutated (ATM) and the nuclear localization signal (NLS) that is required for ATM activation. It also regulates tumor growth partly through ATM signaling. It is a novel tumor suppressor, especially in pancreas and regulates tumor growth potentially through p53 signaling pathway. It also plays a role in stable gene silencing within the nucleus.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Li et al.
Cell cycle (Georgetown, Tex.), 7(18), 2886-2893 (2008-09-13)
Chmp1A (Chromatin modifying protein 1A/Charged multivesicular protein 1A) is a member of the ESCRT-III (Endosomal Sorting Complex Required for Transport) family that was shown to function in endosome-mediated trafficking via multivesicular body (MVB) formation and sorting. Recent reports suggest that
D R Stauffer et al.
Journal of cell science, 114(Pt 13), 2383-2393 (2001-09-18)
The Polycomb-group (PcG) is a diverse set of proteins required for maintenance of gene silencing during development. In a screen for conserved partners of the PcG protein Polycomblike (Pcl), we have identified a new protein, human CHMP1 (CHromatin Modifying Protein;
Zhenqiang You et al.
Cancer letters, 319(2), 190-196 (2012-01-21)
Renal cell carcinoma (RCC) is a highly malignant and often fatal disease of the kidney. Chmp1A is a member of the Endosomal Sorting Complex Required for Transport (ESCRT-III) family, and plays a role in the cytoplasm in sorting proteins to
Sumanth Manohar et al.
Cell cycle (Georgetown, Tex.), 10(15), 2529-2539 (2011-06-28)
Chromatin modifying protein 1A (Chmp1A) is a member of the Endosormal sorting complex required for transport (ESCRT)-III family whose over-expression induces growth inhibition, chromatin condensation, and p53 phosphorylation. p53 is a substrate for Ataxia telangiectasia mutated (ATM), which can be

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service