Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW20081F

Sigma-Aldrich

Anti-Histone H1,1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$418.00

$418.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
50 μG
$418.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$418.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

manufacturer/tradename

Genway 15-288-20081F

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HIST1H1A(3024)

Immunogen

Histone H1.1 native protein.
Immunogen Sequence: GI # 121903, sequence 1-104

Application

Anti-Histone H1,1 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Histone H1.1 is a protein encoded by the HIST1H1A gene in humans and the gene is expressed in testis and thymus, but not in other human tissues. It is located on chromosome 6(6p21-p22) containing 55 histone genes. ist1 on mouse chromosome 13 contains 51 histone genes. There are 2 different human H1 histones coding genes that are differentiated mainly due to the N- and C-terminal H1 domains, whereas the central part of the protein is highly conserved. Histone H1 is a fundamental chromosomal protein and is associated with the maintenance of higher-ordered chromatin organization. It functions in the compaction of chromatin into higher order structures and its modulation is an important step in the potentiation/depotentiation of chromatin structure for transcription. H1 binds less tightly than other histones to DNA in chromatin and can readily exchange in living cells.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Burfeind et al.
Human genetics, 94(6), 633-639 (1994-12-01)
The expression of a human histone H1 isoform (H1.1) was studied in several human tissues. Northern blot analysis has revealed that this gene is expressed in testis and thymus, but not in other human tissues. In this report, we demonstrate
M A Lever et al.
Nature, 408(6814), 873-876 (2000-12-29)
The considerable length of DNA in eukaryotic genomes requires packaging into chromatin to fit inside the small dimensions of the cell nucleus. Histone H1 functions in the compaction of chromatin into higher order structures derived from the repeating 'beads on
S Eick et al.
European journal of cell biology, 49(1), 110-115 (1989-06-01)
The genes coding for two different human H1 histones were isolated, and the primary structures were deduced from the nucleotide sequences. The genes differ from each other and from any other vertebrate H1 structure described until now. The differences occur
Kohsuke Kato et al.
Journal of cell science, 124(Pt 19), 3254-3265 (2011-09-24)
Linker histone H1 is a fundamental chromosomal protein involved in the maintenance of higher-ordered chromatin organization. The exchange dynamics of histone H1 correlates well with chromatin plasticity. A variety of core histone chaperones involved in core histone dynamics has been
William F Marzluff et al.
Genomics, 80(5), 487-498 (2002-11-01)
The multigene family encoding the five classes of replication-dependent histones has been identified from the human and mouse genome sequence. The large cluster of histone genes, HIST1, on human chromosome 6 (6p21-p22) contains 55 histone genes, and Hist1 on mouse

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service