Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW10062F

Sigma-Aldrich

Anti-Somatotropin antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GH, Anti-GHN, Anti-Growth hormone, Anti-Growth hormone 1, Anti-Pituitary growth hormone, Anti-hGH-N

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-10062F

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GH1(2688)

Immunogen

Immunogen Sequence: GI # 13027812, sequence 1-217
Recombinant growth hormone 1 isoform 1

Application

Anti-Somatotropin antibody produced in chicken is suitable for western blotting at a working dilution of 1:500 and for indirect ELISA.

Biochem/physiol Actions

Somatotropin is essential for growth control by stimulating the liver and other tissues to secrete IGF-1. Mutations in this gene causes deficiency of growth hormone and results in dwarfism.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mahalakshmi Santhanam et al.
Molecular endocrinology (Baltimore, Md.), 27(12), 2080-2092 (2013-10-11)
The diverse roles of IGF-1 in physiology include acting as the endocrine intermediate to elicit the anabolic actions of GH. The majority of serum IGF-1 is synthesized in liver, where GH stimulates Igf1 gene transcription via the transcription factor, signal
A new mutation causing inherited growth hormone deficiency: a compound heterozygote of a 6.7 kb deletion and a two base deletion in the third exon of the GH-1 gene.
Y Igarashi et al.
Human molecular genetics, 2(7), 1073-1074 (1993-07-01)
David S Millar et al.
Human mutation, 21(4), 424-440 (2003-03-26)
Subtle mutations in the growth hormone 1 (GH1) gene have been regarded as a comparatively rare cause of short stature. Such lesions were sought in a group of 41 individuals selected for short stature, reduced height velocity, and bone age

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service