Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GS12N0-HH

Sigma-Aldrich

BNPPOC-dG(tac) Phosphoramidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H57N8O12P
Molecular Weight:
968.99
UNSPSC Code:
12352200

type

for DNA synthesis

product line

Proligo Reagents

assay

≥90% (-20 - 200ppm, 31P-NMR)
≥95% (100-200ppm, 31P-NMR)
≥96.0% (reversed phase HPLC)
≥98.0% (3′OH nuclesoside purity, reversed phase HPLC)

packaging

pkg of 12 × 1 g GS12N01-12
pkg of 1 × 10.4 g GS12N10-01
pkg of GS12N00-C (on demand)

impurities

≤0.1 wt. % water content (Karl Fischer)
<0.50% 3′OH nucleside single impurity (reversed phase HPLC)

λ

conforms (UV/VIS Identity)

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service