Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GN952700

Greiner bottle, medium, square

PET, 1000 mL, sterile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

material

PET bottle
clear bottle
narrow-mouth bottle
square bottle

feature

autoclavable: no
cap

packaging

pack of 24 ea

manufacturer/tradename

Greiner 952700

availability

available only in China

capacity

500 mL

Looking for similar products? Visit Product Comparison Guide

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service