Key Documents
GN781270
Greiner MASTERBLOCK® storage microplate
Deep Well, 384 well, polypropylene, capacity 240 uL, conical (V) bottom, translucent
Synonym(s):
384 well microplate, 384 well plate, 384 well storage microplate, cell culture plate
About This Item
UNSPSC Code:
41121800
material:
polypropylene
size:
384 wells
Pricing and availability is not currently available.
material
polypropylene
packaging
pack of 60 ea
manufacturer/tradename
Greiner 781270
availability
available only in China
size
384 wells
suitability
suitable for (general storage, liquid handling; automated applications)
Features and Benefits
  • Detectable DNase, RNase, human DNA free
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Can be tracked back to manufacturing using a LOT numbering system.
  • Allows for a wide range of applications when higher volumes are required.
  • In general, it holds good for compound libraries and the storage of samples
  • The wells′ conical shape allows for accurate pipetting down to the last drop.
  • The standardized external dimensions and the tight tolerances facilitate easy liquid handling for robotics
  • Has solid V-bottom (conical bottom) and alphanumeric well coding
  • Offers high chemical and temperature (−196°C to +121°C) resistance
  • Sealable with CapMats and adhesive films and heat sealer
Legal Information
MASTERBLOCK is a registered trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)