Key Documents
GN780276
Greiner MASTERBLOCK® storage microplate
96 well, polypropylene, capacity 2.0 mL, conical (V) bottom, yellow
About This Item
UNSPSC Code:
41121800
material:
polypropylene
size:
96 wells
material
polypropylene
packaging
pack of 50 ea
manufacturer/tradename
Greiner 780276
availability
available only in China
size
96 wells
suitability
suitable for (general storage, liquid handling; automated applications)
Legal Information
MASTERBLOCK is a registered trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)