Key Documents
GN780201
Greiner MASTERBLOCK® storage microplate
96 well, polypropylene, capacity 1.0 mL, round (U) bottom, translucent
About This Item
UNSPSC Code:
41121800
material:
polypropylene
size:
96 wells
Pricing and availability is not currently available.
material
polypropylene
packaging
pack of 50 ea
manufacturer/tradename
Greiner 780201
availability
available only in China
size
96 wells
suitability
suitable for (general storage, liquid handling; automated applications)
Legal Information
MASTERBLOCK is a registered trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)