Key Documents
GN755001
Greiner Multiwell Strips
medium binding, round bottom (16 well, 2 × 8 strips), non-sterile
Synonym(s):
Greiner Multiwell Strips
About This Item
UNSPSC Code:
41121800
size:
16 well
sterility:
non-sterile
binding type:
medium binding surface
sterility
non-sterile
packaging
pack of 300 ea
manufacturer/tradename
Greiner 755001
availability
available only in China
size
16 well
binding type
medium binding surface
General description
Both medium-binding and high binding are offered. High binding plates and strips are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding are suggested.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)