Key Documents
GN754074
Greiner microplate
2 x 8 well, U-strip, high binding, white
Synonym(s):
2 x 8 well microplate, 2 x 8 well plate, cell culture plate
About This Item
UNSPSC Code:
41121800
material:
white
material
white
packaging
pack of 100 ea
manufacturer/tradename
Greiner 754074
availability
available only in China
suitability
suitable for (cell culture)
General description
  • Greiner microplates are ideal for storing active agents, patient samples, DNA/RNA or stock cultures.
  • Polypropylene material can withstand a high temperature (-196 to +121°C) and are chemically resistant.
  • Nonpyrogenic plates are free of detectable DNase, RNase, human DNA.
  • Features alphanumeric well coding for identification.
  • Sealable with adhesive films and heat sealers for long-term and secure sealing.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)