Key Documents
GN754070
Greiner MICROLON® 200 strip microplate
6 x U16, 96 well, round U-bottom, medium binding, clear
About This Item
UNSPSC Code:
41121800
material:
clear
size:
96 wells
Pricing and availability is not currently available.
material
clear
packaging
pack of 100 ea
manufacturer/tradename
Greiner 754070
availability
available only in China
size
96 wells
suitability
suitable for (cell culture)
Legal Information
MICROLON is a registered trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)