Key Documents
GN676090
Greiner SILVERseal sealer, adhesive aluminium foil on carrier paper, piercable, silver
adhesive aluminium foil on carrier paper, piercable, silver
About This Item
UNSPSC Code:
41122410
Pricing and availability is not currently available.
material
acrylic (Glue:adhesive)
aluminum foil
sterility
non-sterile
feature
piercable
packaging
pack of 100 ea
manufacturer/tradename
Greiner 676090
availability
available only in China
Legal Information
SILVERseal is a trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)