Key Documents
GN676061
Greiner plate sealer
non-toxic, sterile
About This Item
UNSPSC Code:
41122410
material
acrylic (Glue: DURO AL 170 (solvent))
foil (PET (Polyethyleneterephtalat))
paper (Silicone-paper KV 75)
sterility
sterile
packaging
pack of 100 ea
manufacturer/tradename
Greiner 676061
availability
available only in China
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)