Key Documents
GN675075
Greiner half-area microplate
96 well, flat bottom, medium binding, white
About This Item
UNSPSC Code:
41121800
material:
white
size:
96 wells
Pricing and availability is not currently available.
material
white
packaging
pack of 40 ea
manufacturer/tradename
Greiner 675075
availability
available only in China
size
96 wells
suitability
suitable for (cell culture)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)