Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GN671277

Greiner Sapphire PCR tubes, thin wall

volume 0.2 mL, attached dome, violet

Synonym(s):

Greiner PCR tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
Pricing and availability is not currently available.

material

polypropylene

packaging

pack of 10000 ea

manufacturer/tradename

Greiner 671277

availability

available only in China

technique(s)

PCR: suitable

suitability

suitable for (PCR applications)

Looking for similar products? Visit Product Comparison Guide

General description

Greiner PCR tubes′ thin wall construction promotes optimal heat transfer between the thermocycler block and the reaction mixture. These PCR tubes are certified RNase/DNase free and are non-pyrogenic as well.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service