Key Documents
GN661160
Greiner CELLSTAR® T-175 flask
TC treated, 650 mL, HiProfile, w/ standard cap
Synonym(s):
cell culture flask, culture flask, standard cap cell culture flask, treated flask
About This Item
UNSPSC Code:
41122104
Pricing and availability is not currently available.
packaging
pack of 40 ea
manufacturer/tradename
Greiner 661160
availability
available only in China
Legal Information
CELLSTAR is a registered trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)