Key Documents
GN660175
Greiner CELLSTAR® T-175 flask
TC treated, 550 mL, LoProfile, w/ filter cap
Synonym(s):
cell culture flask, culture flask, filter cap cell culture flask, treated flask
About This Item
UNSPSC Code:
41122104
Pricing and availability is not currently available.
packaging
pack of 50 ea
manufacturer/tradename
Greiner 660175
availability
available only in China
General description
  • Greiner CELLSTAR® T-175 culture flasks are made from high-quality polystyrene.
  • Flasks are non-pyrogenic, non-cytotoxic, and free of detectable DNase, RNase, and human DNA.
  • Canted neck design facilitates easy access to samples with a cell scraper or pipette.
  • High-profile neck ensures additional protection against contamination.
  • TC (tissue-culture) surface treatment offers optimal conditions for improved maintenance of adherent cells.
  • Graduation on the side for easy filling.
  • Features a red filter screw cap for sample protection.
Legal Information
CELLSTAR is a registered trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)