Key Documents
GN655096
Greiner FLUOTRAC 200 microplate
96 well, flat μClear® bottom, medium binding, chimney style, black
About This Item
UNSPSC Code:
41121800
material:
black
size:
96 wells
Pricing and availability is not currently available.
material
black
packaging
pack of 40 ea
manufacturer/tradename
Greiner 655096
availability
available only in China
size
96 wells
suitability
suitable for (cell culture)
Legal Information
μClear is a registered trademark of Greiner Bio-One GmbH
FLUOTRAC is a trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)