Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GN654180

Greiner HLA Terasaki multiwell plates

size 72 wells, 1 lid per plate

Synonym(s):

72 well microtest plate, Greiner HLA Terasaki multiwell plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
size:
72 wells
Pricing and availability is not currently available.

packaging

pack of 270 ea

manufacturer/tradename

Greiner 654180

availability

available only in China

size

72 wells

well size

10 μL

suitability

suitable for (cell culture)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

HLA Microtest plates (Terasaki) from Greiner. Designed for tissue typing techniques to ensure smooth spreading of sera and cells across the flat base of the well. The surface of each well gives a microscopically precise and clear picture. All plates are stackable.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service