Key Documents
GN650162
Greiner microplate
96 well, round bottom, clear
Synonym(s):
96 well microplate, 96 well plate, cell culture plate
About This Item
UNSPSC Code:
41121800
material:
clear
size:
96 wells
Pricing and availability is not currently available.
material
clear
packaging
pack of 375 ea
manufacturer/tradename
Greiner 650162
availability
available only in China
size
96 wells
suitability
suitable for (cell culture)
General description
  • Greiner microplates are ideal for storing active agents, patient samples, DNA/RNA or stock cultures.
  • Polypropylene material can withstand a high temperature (-196 to +121°C) and are chemically resistant.
  • Nonpyrogenic plates are free of detectable DNase, RNase, human DNA.
  • Features alphanumeric well coding for identification.
  • Sealable with adhesive films and heat sealers for long-term and secure sealing.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)