Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GN541080

Greiner cell scrapers

40 cm handle

Synonym(s):

cell lifter, cell lifters, cell scraper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122103
Pricing and availability is not currently available.

sterility

sterile

manufacturer/tradename

Greiner 541080

availability

available only in China

Looking for similar products? Visit Product Comparison Guide

General description

Greiner cell scrapers are an ideal combination of a rigid polystyrene handle with a soft polyethylene scraping blade.

Material
Handle: Polystyrene
Blade: High Density Polyethylene

Color
Handle: Blue
Blade: White

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service