Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GN188283

Greiner centrifuge tubes

volume 15 mL, 17x120 mm, blue cap, conical bottom, light protection

Synonym(s):

centrifuge tubes, culture tubes, plastic centrifuge tube, plastic culture tube, Greiner centrifuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
Pricing and availability is not currently available.

material

polypropylene tube

sterility

sterile

packaging

pack of 500 ea

manufacturer/tradename

Greiner 188283

availability

available only in China

color

colorless tube

Looking for similar products? Visit Product Comparison Guide

General description

Greiner centrifuge tubes
High quality, crystal-clear polypropylene centrifuge tubes. Centrifuge tubes can be safely centrifuged to approximately 3,200 × g (50 mL size) and 3,500 × g (15 mL size) depending on the specific gravity of the solution being centrifuged and the type of rotor.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service