GEUS79010
ExoProStar™ S 100
Cytiva US79010, 100 reactions
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106300
NACRES:
NA.52
usage
100 reactions
manufacturer/tradename
Cytiva US79010
availability
not available in USA, Canada and Puerto Rico
shipped in
dry ice
storage temp.
−20°C
General description
ExoProStar S 100 rxn is an Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs in 15 minutes. For maximum flexibility it comes in two separate tubes.
Application
The ExoProStar S PCR and Sequence Reaction Clean-Up Kit updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs. illustra™ ExoProStar S improves digestion efficiency with no degradation of the target PCR product. Your sample is completely intact and ready for immediate use in manual or automated processes.
Features and Benefits
- Easy to automate
- Enzymes provided in two separate tubes; just two simple pipetting steps are needed to prepare the reaction
- Enzymes optimized to work together for high efficiency removal of unincorporated primers and nucleotides
- Scalable for different reaction sizes
- No loss of PCR product
- Complete heat inactivation of the enzymes within 10 min
- Fast 15 min protocol
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
ExoProStar is a trademark of Cytiva
illustra is a trademark of Cytiva
Storage Class
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service