Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GEUS79002

ExoProStar S 20

Cytiva US79002, 20 reactions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.52

usage

20 reactions

manufacturer/tradename

Cytiva US79002

availability

not available in USA, Canada and Puerto Rico

shipped in

dry ice

storage temp.

−20°C

General description

ExoProStar S 20 rxn is an Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs in 15 minutes. For maximum flexibility it comes in two separate tubes.

Application

The ExoProStar S PCR and Sequence Reaction Clean-Up Kit updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs. illustra ExoProStar S improves digestion efficiency with no degradation of the target PCR product. Your sample is completely intact and ready for immediate use in manual or automated processes.

Features and Benefits

  • Easy to automate
  • Enzymes provided in two separate tubes; just two simple pipetting steps are needed to prepare the reaction
  • Enzymes optimized to work together for high efficiency removal of unincorporated primers and nucleotides
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Complete heat inactivation of the enzymes within 10 min
  • Fast 15 min protocol

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

ExoProStar is a trademark of Cytiva
illustra is a trademark of Cytiva

Storage Class

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service