GETE62
GE TE62 Transphor transfer unit
AC/DC input 100 V AC
All Photos(1)
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Cytiva Article No. 80-6209-58
Cytiva TE62
General description
TE 62 Transfer Cooled Unit
The gel cassettes provide a stiff backbone for the gel and membrane sandwiches. The hinged halves lock together for equal pressure across the gel. The grid design of the gel cassettes maximizes the open area for even transfers of gels up to 15 × 21 cm. The crucial element for uniform electrophoretic transfer, a uniform electrical field, is assured by spacing the electrode panels 10 cm apart and incorporating 140 cm of wire in an evenly-spaced pattern. For safety, the electrode panels are locked securely into the Transfer Unit, but can be easily removed for cleaning or replacement.
Runs up to 4, 15 x 21 cm gels or 17, 7 x 10 cm mini gels.
- New hinged cassettes molded of tough, glass-filled polyethersulfone for increased durability.
- Transfer four standard gels or 16 mini-gels.
- Load cassettes easily with even contact between the gel and transfer membrane.
- Secure, sturdy electrode panels lock automatically in place.
- Efficient transfer due to evenly spaced vertical platinum wire on electrode panel.
The gel cassettes provide a stiff backbone for the gel and membrane sandwiches. The hinged halves lock together for equal pressure across the gel. The grid design of the gel cassettes maximizes the open area for even transfers of gels up to 15 × 21 cm. The crucial element for uniform electrophoretic transfer, a uniform electrical field, is assured by spacing the electrode panels 10 cm apart and incorporating 140 cm of wire in an evenly-spaced pattern. For safety, the electrode panels are locked securely into the Transfer Unit, but can be easily removed for cleaning or replacement.
Runs up to 4, 15 x 21 cm gels or 17, 7 x 10 cm mini gels.
Choose from one of the most recent versions:
Certificates of Analysis (COA)