Key Documents
GESE26010A75
GE SE260 mini-vertical unit for two slab gels
complete, gel size 10 cm × 10.5 cm, AC/DC input 500 V AC
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
packaging
pack of 1 ea
manufacturer/tradename
Cytiva Article No. 80-6149-35
Cytiva SE260-10A-,75
gel size
10 cm × 10.5 cm
General description
Offers the combined advantages of small format and efficient cooling, an ideal combination for rapid screening of protein and nucleic acid samples. Gels are efficiently cooled using the heat-conductive alumina gel plate and built in heat-exchanger, with or without connection to an external circulator. The alumina back plate of each gel sandwich is in contact with buffer in one of the two buffer chambers, which act as heat sinks. The alumina plate conducts heat away from the gel sandwich into the upper buffer at a rate 40-times faster than glass of comparable thickness. In addition, a serpentine channel through which coolant can be pumped is permanently sealed between the two upper buffer chambers
Runs two 10 cm × 10.5 cm gels, the additional 2.5 cm of length gives more than 25% extra distance to resolve complex samples,you can use a variety of 8 and 10 cm long precast gels.
Choose from one of the most recent versions:
Certificates of Analysis (COA)