Key Documents
GESE215
GE Mighty Small multiple gel caster
for use with SE250, 5 to 10 gels, 10 x 8 cm plates
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
packaging
pack of 1 ea
manufacturer/tradename
Cytiva Article No. 80-6142-51
Cytiva SE215
Choose from one of the most recent versions:
Certificates of Analysis (COA)